Learn More
Immunology Consultants anti-Dog IgM Antibody (Goat) - HRP Conjugated
SDP
Catalog No. 502248896
Encompass_Preferred
This HRP‑conjugated goat antibody enables direct, enzyme‑based detection of canine IgM, providing strong chemiluminescent or colorimetric signal in ELISA and Western blot formats. The conjugate maintains the specificity of the affinity‑purified parent antibody while streamlining workflows by eliminating the need for secondary reagents. GM‑40P is ideal for high‑sensitivity veterinary assays, infection monitoring, and studies requiring rapid, robust IgM detection.
This item is not returnable. View return policy
Catalog No. 50-224-8896 Supplier Immunology Consultants Supplier No. GM40P
This HRP‑conjugated goat antibody enables direct, enzyme‑based detection of canine IgM, providing strong chemiluminescent or colorimetric signal in ELISA and Western blot formats. The conjugate maintains the specificity of the affinity‑purified parent antibody while streamlining workflows by eliminating the need for secondary reagents. GM‑40P is ideal for high‑sensitivity veterinary assays, infection monitoring, and studies requiring rapid, robust IgM detection.