Learn More
Invitrogen™ Goat anti-Donkey IgG (H+L) Secondary Antibody, Texas Red
Shop All Thermo Scientific ProductsDescription
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Donkey IgG (H+L) |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Texas Red |
| Formulation | PBS with 1% BSA and 0.01% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Donkey IgG whole molecule. |
| Purification Method | IgG fraction |
| Quantity | 1 mg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.