Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Guinea Pig, AMCA, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ108MMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ108MMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ108MMI Supplier MilliporeSigma Supplier No. AQ108M
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Guinea Pig IgG using this Goat anti-Guinea Pig IgG Antibody, F(ab′) 2, AMCA conjugate validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Guinea Pig IgG
Applications Immunofluorescence
Classification Polyclonal
Conjugate AMCA
Host Species Goat
Quantity 1 mg
Primary or Secondary Secondary
Target Species Guinea Pig

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.