Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Guinea Pig, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ108PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ108PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ108PMI Supplier MilliporeSigma Supplier No. AQ108P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Detect Guinea Pig IgG using this Goat anti-Guinea Pig IgG Antibody, F(ab′) 2, HRP validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Guinea Pig IgG F(ab')2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Host Species Goat
Quantity 500 μL
Primary or Secondary Secondary
Target Species Guinea Pig

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.