Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Guinea Pig IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 680

Catalog No. PISA510098 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PISA510098 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PISA510098 Supplier Invitrogen™ Supplier No. SA510098
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Secondary Antibody

This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, horse, human, mouse, rabbit, rat and sheep.

Anti-Guinea Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for guinea pig immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Guinea Pig IgG (H+L) Cross-Adsorbed, Guinea pig IgG (H+L)
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate DyLight 680
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Host Species Goat
Immunogen Guinea pig IgG-heavy and light chain.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Guinea Pig
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.