Learn More
Immunology Consultants anti-Hamster IgG h+l (min. reactivity w/ Ms, Rt) - Affinity Purified
SDP
Catalog No. 502248877
Encompass_Preferred
This affinity‑purified goat antibody provides broad, sensitive detection of hamster IgG by recognizing both heavy and light chains while minimizing cross‑reactivity with mouse and rat immunoglobulins. The reduced off‑target binding improves assay clarity in ELISA, Western blotting, immunofluorescence, and other secondary detection workflows involving rodent models. Its strong affinity and low background support reliable detection of hamster‑derived primary antibodies in complex biological samples. GHHL‑65AX9025 is well suited for multiplex studies, CHO‑based research, and applications requiring species‑selective secondary detection with consistent, reproducible performance.
This item is not returnable. View return policy
Catalog No. 50-224-8877 Supplier Immunology Consultants Supplier No. GHHL65AX9025
This affinity‑purified goat antibody provides broad, sensitive detection of hamster IgG by recognizing both heavy and light chains while minimizing cross‑reactivity with mouse and rat immunoglobulins. The reduced off‑target binding improves assay clarity in ELISA, Western blotting, immunofluorescence, and other secondary detection workflows involving rodent models. Its strong affinity and low background support reliable detection of hamster‑derived primary antibodies in complex biological samples. GHHL‑65AX9025 is well suited for multiplex studies, CHO‑based research, and applications requiring species‑selective secondary detection with consistent, reproducible performance.