Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-, HRP, Clone: , Southern Biotech™
SDP

Catalog No. OB530005B Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB530005B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB530005B Supplier Southern Biotechnology Associates Supplier No. 530005B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Secondary Antibody

SouthernBiotech Clonotyping kits are designed for the isotyping of mouse monoclonal antibodies and may also be used for quantitative studies.

Specifications

Applications ELISA
Conjugate HRP
Formulation UNLB antibody - Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added HRP conjugates - Supplied in a stock solution in 50% glycerol/50% PBS, pH 7.4; No preservative added
Host Species Goat
Quantity 1 Kit
Regulatory Status RUO
Primary or Secondary Secondary
Content And Storage Store at 2-8°C; Protect from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.