Learn More
Immunology Consultants Affinity Purified Goat anti-Human C4 Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants GC480A
Encompass_Preferred
Goats were immunized with highly purified human C4 and the resulting antiserum was collected. Antibody was immunoaffinity purified off a C4 containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human C4 as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA, and IHC applications. This antibody may cross-react with C4 from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8793