Learn More
Immunology Consultants anti-Human Fibronectin Antibody (Goat) - Antiserum
SDP
Catalog No. 502248827
Encompass_Preferred
This goat antiserum provides broad, polyclonal detection of human fibronectin, a major extracellular matrix glycoprotein involved in adhesion, migration, and tissue remodeling. The antiserum supports ELISA, immunodiffusion, and other applications requiring comprehensive epitope recognition. GFCT‑80 is well suited for cell biology, wound‑healing research, and studies examining matrix‑associated signaling pathways. Its broad reactivity ensures reliable detection across purified proteins, cell lysates, and complex biological samples.
This item is not returnable. View return policy
Catalog No. 50-224-8827 Supplier Immunology Consultants Supplier No. GFCT80
This goat antiserum provides broad, polyclonal detection of human fibronectin, a major extracellular matrix glycoprotein involved in adhesion, migration, and tissue remodeling. The antiserum supports ELISA, immunodiffusion, and other applications requiring comprehensive epitope recognition. GFCT‑80 is well suited for cell biology, wound‑healing research, and studies examining matrix‑associated signaling pathways. Its broad reactivity ensures reliable detection across purified proteins, cell lysates, and complex biological samples.