Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig κ chain Goat anti-Human, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP505AMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP505AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP505AMI Supplier MilliporeSigma Supplier No. AP505A
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Human Ig κ chain using this Goat anti-Human Ig κ chain Antibody, Alkaline Phosphatase conjugate validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human Ig κ chain
Applications ELISA
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Purified by affinity chromatography on pooled human Igs with kappa light chains covalently linked to agarose. Liquid in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative.
Host Species Goat
Immunogen pooled antisera from goats hyperimmunized with human myeloma proteins with kappa light chains.
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C under sterile conditions for up to 12 months −20°C in undiluted aliquots for long term storage. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.