Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig Goat anti-Human, BGAL (β-galactosidase), Polyclonal, Southern Biotech™
SDP

Catalog No. OB201006 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
1.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB201006 1.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB201006 Supplier Southern Biotechnology Associates Supplier No. 201006
Add to Cart
Add to Cart

Goat Polyclonal Secondary Antibody

SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.

Specifications

Antigen Ig
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Lot specific
Conjugate b-GAL (beta-Galactosidase)
Formulation Supplied in a stock solution in 50% glycerol/50% PBS containing NaN3 as preservative
Host Species Goat
Immunogen Human IgG, IgM, and IgA
Purification Method Affinity Purified
Quantity 1.0 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 2-8°C
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.