Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Human Ig lambda; chain, Rhodamine (TRITC), Secondary Antibody,

Catalog No. AP506RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
Conjugate:
Rhodamine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AP506RMI 1 mg Rhodamine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP506RMI Supplier MilliporeSigma™ Supplier No. AP506R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Specifically detects Ig λ chain Clone: in Human samples, and it is validated for ELISA, Immunofluorescence

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and lambda light chains are approximately 250 amino acids in length with an average mass of about 25kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig λ chain
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Rhodamine
Formulation Purified by affinity chromatography on pooled human Igs with lambda light chains covalently linked to agarose. Liquid in 1.0 mL PBS/NaN3.
Host Species Goat
Immunogen Prepared from pooled antisera from goats hyperimmunized with human myeloma proteins with lambda light chains.
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Secondary, Control Antibodies
Primary or Secondary Secondary
Test Specificity The antibody reacts with human lambda light chains as demonstrated by ELISA and flow cytometry.
Target Species Human
Content And Storage 2°C to 8°C under sterile conditions for up to twelve months from date of receipt; Do not freeze
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.