Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Human IgA, alpha-Chain Specific, Alkaline Phosphatase, Secondary Antibody,

Catalog No. 4011321ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
Conjugate:
Alkaline Phosphatase (AP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-113-21ML 1 mL Alkaline Phosphatase (AP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-113-21ML Supplier MilliporeSigma™ Supplier No. 4011321ML
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects IgA, α-Chain Specific Clone: in Human samples, and it is validated for Enzyme Immunoassay, Immunoelectrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA, α-Chain Specific
Applications ELISA, Immunoelectrophoresis
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation In 50mM Tris, 1mM MgCl2, 0.1mM ZnCl2, 1% BSA, pH 7.6.
Host Species Goat
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 2°C to 8°C
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.