Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgA alpha heavy chain F(ab')2 fragment, HRP, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89138415 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-415 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-415 Supplier Abnova Corporation Supplier No. PAB25510
Add to Cart
Add to Cart

Specifications

Antigen Goat anti-Human IgA alpha heavy chain F(ab')2 fragment
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Description Goat anti-human IgG recognizes human IgA alpha heavy chain F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography.
Dilution ELISA (1:5000-1:100000) Western Blot (1:5000-1:100000) Immunochemistry (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM KH2PO4, 250mM NaCl, pH 7.2 (15mg/mL BSA. Do NOT add Sodium Azide!)
Host Species Goat
Immunogen Human IgA alpha heavy chain F(ab')2 fragment.
Purification Method This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation.
Quantity 500 μL
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 0.5mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.