Learn More
Invitrogen™ Goat anti-Human IgA (Heavy chain) Secondary Antibody
Shop All Thermo Scientific ProductsDescription
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31140 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31140 reacts with the heavy chain portion of human IgA, but not with the heavy chains of human IgG or IgM. The antibody does not react against the light chains common to most human immunoglobulins, or against non-immunoglobulin serum proteins. However, this antibody may cross-react with IgA from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of Origin: USA.
Specifications
Specifications
| Antigen | Human IgA (Heavy chain) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 2 mg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.