Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA Goat anti-Human, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP120RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP120RMI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP120RMI Supplier MilliporeSigma Supplier No. AP120R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Goat anti-Human IgA Antibody, IgG and IgM, Rhodamine conjugate is an antibody against Human IgA for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgA IgG and IgM
Applications Immunofluorescence
Classification Polyclonal
Conjugate Rhodamine
Host Species Goat
Quantity 2 mg
Primary or Secondary Secondary
Target Species Human

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.