Learn More
Immunology Consultants Biotin Conjugated Goat anti-Human IgE Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants GE80B
Encompass_Preferred
Goats were immunized with highly purified human IgE and the resulting antiserum was collected. Antibody was solid phase adsorbed, immunoaffinity purified off a human IgE containing immunosorbent and conjugated with biotin. Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgE as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA, and IHC applications. This antibody may cross-react with IgE from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2183