Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Human IgE, epsilon-Chain Specific, Secondary Antibody,

Catalog No. 4115201ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
41-152-01ML 1 mL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-152-01ML Supplier MilliporeSigma™ Supplier No. 4115201ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Specifically detects IgE, ε-Chain Specific Clone: in Human samples, and it is validated for Immunoelectrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgE, ε-Chain Specific
Applications Immunoelectrophoresis
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum diluted in 500mM NaCl, 50mM Tris-HCl, pH 7.5.
Host Species Goat
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage −20°C
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.