Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Human, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP112AMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP112AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP112AMI Supplier MilliporeSigma Supplier No. AP112A
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Human IgG using this Goat anti-Human IgG Antibody, Alkaline Phosphatase conjugate validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Lyophilized. Buffer = 0.01M Tirs HCl, 0.25M NaCl, pH 8.0 with 15mg/mL BSA and 0.05% sodium azide. RECONSTITUTION: Reconstitute with sterile distilled water to match the volume indicated on the vial label.
Host Species Goat
Purification Method Serum
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.