Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG γ chain Goat anti-Human, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP504BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP504BMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP504BMI Supplier MilliporeSigma Supplier No. AP504B
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Goat anti-Human IgG γ chain Antibody, biotin conjugate, Species Adsorbed is validated for use in ELISA, immunofluorescence for the detection of Human IgG γ chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgG γ chain Species adsorbed
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation Purified by affinity chromatography on pooled human IgG covalently linked to agarose. Liquid in 1.0mLPBS/NaN3.
Host Species Goat
Immunogen IgG γ chain. Pooled antisera from goats hyperimmunized with normal human IgG and human IgG paraproteins.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.