Learn More
Bio Assay Works Dressed GoldGoat anti-Human IgG (H+L) Conjugates, 9ml, 15 O.D./ml
Supplier:  Bio Assay Works DGHGB009
Encompass
Dressed Gold Goat anti-Human IgG (H+L) conjugates are produced using Goat anti-Human IgG (H+L) polyclonal antibodies and BioAssay Works proprietary 40 nmNaked Goldsols. The conjugates are highly reactive and ideal for use in lateral flow assays where high sensitivity is required. Specifications:O.D./mL: 15 or 50Storage temperature: 2-8 C, DO NOT FREEZE!Shipping temperature: 2-8 CStability: 2 years from date of manufacture
Catalog No. NC1481179