Learn More
Invitrogen™ Goat anti-Human IgG F(ab')2 Cross-Adsorbed Secondary Antibody
Shop All Thermo Scientific ProductsDescription
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31132 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31132 reacts with the light chains of human IgG and with the light chains common to other human immunoglobulins, but not with the Fc portion of human IgG or non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, horse and mouse serum proteins. However, the antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of origin: USA.
Specifications
Specifications
| Antigen | Human IgG F(ab')2 Cross-Adsorbed |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.79 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1.5 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.