Learn More
Goat anti-Human IgG F(ab')2 fragment, FITC, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Goat anti-Human IgG F(ab')2 fragment |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Description | Goat anti-human IgG recognizes human IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide), 10mg/mL BSA (Immunoglobulin and Protease free). Restore with 1mL deionized water (or equivalent) |
| Host Species | Goat |
| Immunogen | Human IgG F(ab')2 fragment |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.