Learn More
Invitrogen™ Goat anti-Human IgG F(ab')2 Secondary Antibody, FITC
Shop All Thermo Scientific ProductsDescription
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31628 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31628 reacts with F(ab')2 /Fab portion of human IgG and with the light chains common to other human immunoglobulins, but does not react against the Fc protion of human IgG or against non-immunoglobulin serum proteins. The antibody exhibits minimal cross-reactivity to mouse serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 8.2 μg/mL; > 3 moles FITC per mole IgG (lot-dependent). Reconstitute with 1.5 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA.
Specifications
Specifications
| Antigen | Human IgG F(ab')2 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Host Species | Goat |
| Purification Method | Affinity chromatography |
| Quantity | 2 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.