Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Human, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP113AMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP113AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP113AMI Supplier MilliporeSigma Supplier No. AP113A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Goat anti-Human IgG Antibody, flow cytometry, Alkaline Phosphatase conjugate detects level of Human IgG and has been published and validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Goat secondary antibody IgG in Lyophilized Buffer containing 0.01M Tris HCl, 0.25M NaCl, pH 8.0 with 15mg/mL BSA and 0.05% sodium azide. RECONSTITUTION:
Reconstitute with sterile distilled water to match the volume indicated on the vial label.
Host Species Goat
Purification Method Unpurified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C for up to 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.