Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP113MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP113MI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP113MI Supplier MilliporeSigma Supplier No. AP113
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Goat anti-Human IgG Antibody, Fc is validated for use in ELISA, immunoprecipitation, western blotting for the detection of Human IgG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgG
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2mg/mL
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M Sodium Chloride with 0.1% Sodium Azide.
Host Species Goat
Immunogen Human IgG, Fc
Purification Method Affinity Purified
Quantity 2 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C. Do not freeze.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.