Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Human, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP112FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP112FMI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP112FMI Supplier MilliporeSigma Supplier No. AP112F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Goat anti-Human IgG Antibody, FITC conjugate is an antibody against Human IgG for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgG
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation Lyophilized from 0.02M phosphate buffered saline, 0.25M sodium chloride, pH 7.6, with 0.1% sodium azide and 15mg/mL bovine serum albumin. Reconstitute with 2mLof sterile distilled water. FLUOROPHORE/PROTEIN: Approximately 14μg/mg
Host Species Goat
Immunogen Purified human IgG.
Purification Method Affinity Purified
Quantity 2 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.