Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Human IgG (H+L) Superclonal™ Secondary Antibody, PE

Catalog No. A56015PE
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A56015PE 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A56015PE Supplier Invitrogen™ Supplier No. A56015PE
Only null left
Add to Cart
Add to Cart

Goat Recombinant Superclonal Secondary Antibody

Goat anti-Human IgG (H+L) Superclonal™ Recombinant Secondary Antibody, PE is specific to Human IgG with minimum cross-reactivity towards Guinea pig IgG, Rat IgG, Horse IgG and Monkey IgM. Moderate cross-reactivity was observed with Monkey IgG. Superclonal™ recombinant secondary antibodies are produced using a multifaceted clonal selection process that involves several phenotypic screens to ensure enhanced specificity, animal origin free formulation and lot-to-lot consistency. R-phycoerythrin (R-PE) is an intensely bright phycobiliprotein isolated from red algae that exhibits extremely bright red-orange fluorescence with high quantum yields. In practical applications, the sensitivity of R-PE conjugates is usually 5 to 10 times greater than those of the corresponding fluorescein conjugates. R-PE is a large molecule used for fluorescence-based detection, primarily in flow cytometry, microarray assays, ELISAs, and other applications that require high sensitivity, but not photostability. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG (H+L)
Applications Flow Cytometry
Classification Recombinant Superclonal
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 5mM sodium azide; pH 7.4
Host Species Goat
Immunogen Purified Human IgG.
Purification Method Affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.