Learn More
Goat anti-Human IgG, IgA, IgM whole molecules, HRP, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Goat anti-Human IgG, IgA, IgM whole molecules |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Description | Goat anti-human IgG recognizes human IgG, IgA and IgM whole molecules. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% gentamicLyophilized from sulfate) |
| Host Species | Goat |
| Immunogen | Human IgG, IgA and IgM whole molecules |
| Purification Method | This product was prepared from polyspecific antiserum by immunoaffinity chromatography using antigens coupled to agarose beads. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.