Learn More
Invitrogen™ Goat anti-Human IgG, IgM, IgA (H+L) Secondary Antibody, TRITC
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP). Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. 1 year from date of receipt. Prepare working dilution prior to use and then discard. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin human serum immunoglobulins.
Specifications
Specifications
| Antigen | Human IgG, IgM, IgA (H+L) |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | TRITC |
| Formulation | PBS with 1% BSA and 0.05% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Human IgG/IgA/IgM. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.