Learn More
Invitrogen™ Goat anti-Human IgG, IgM, IgA (H+L) Secondary Antibody
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Specifications
Specifications
| Antigen | Human IgG, IgM, IgA (H+L) |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | >4.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Human IgG/IgA/IgM. |
| Purification Method | Affinity chromatography |
| Quantity | 2 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.