Learn More
Invitrogen™ Goat anti-Human Lambda Light Chain Secondary Antibody
Shop All Thermo Scientific ProductsDescription
This antibody may cross react with lambda light chains from other species.
Specifications
Specifications
| Antigen | Human Lambda Light Chain |
| Applications | ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Human Lambda b+f. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.