Learn More
MyBioSource Goat Anti-Human IgG, Monkey Ads
SDP
Supplier:  MyBioSource MBS6751171 MG
Encompass_Preferred
This product is a secondary antibody from goat that targets human IgG in monkey samples. It is an affinity purified antibody that has been adsorbed against human serum proteins to minimize cross-reactivity. The immunogen is human IgG whole molecule. This antibody can be used in ELISA, WB, IHC, and other immunoassay applications. It is supplied as a lyophilized powder with BSA and sodium azide as stabilizers. Reconstitute with 0.5 mL of PBS.
Catalog No. NC0672104