Learn More
IgG Goat anti-Human, Biotin, Polyclonal, Southern Biotech™
Shop All Southern Biotechnology Associates Products

Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
Specifications
| Antigen | IgG |
| Applications | ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Formulation | Supplied in PBS/NaN3 |
| Host Species | Goat |
| Immunogen | Human IgG |
| Purification Method | Affinity Purified |
| Quantity | 1.0 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.