Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP112MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP112MI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP112MI Supplier MilliporeSigma Supplier No. AP112
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Goat anti-Human IgG Antibody is an antibody against Human IgG for use in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgG Antibody
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2mg/mL
Conjugate Unconjugated
Formulation Provided at 100 μg/100 μL in PBS, pH 7.4, containing 0.2% sodium merthiolate as a preservative.
Host Species Goat
Immunogen Highly purified Human IgG isolated from pooled normal human serum.
Quantity 2 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C for up to 6 months.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.