Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM+IgG+IgA Goat anti-Human, Phycoerythrin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ503HMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ503HMI 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ503HMI Supplier MilliporeSigma Supplier No. AQ503H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Detect Human IgM+IgG+IgA using this Goat anti-Human IgM+IgG+IgA Antibody, F(ab′)2, PE conjugate validated for use in ELISA and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgM IgG IgA
Applications ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.25mg/mL
Conjugate PE
Formulation Purified by gel filtration chromatography of pepsin digested antibody. Liquid in 1.0mLPBS/NaN3.
Host Species Goat
Immunogen pepsin digest of goat anti-human Ig.
Purification Method Gel filtration chromatography
Quantity 0.25 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C under sterile conditions for up to 12 months. Avoid repeated freeze/thaw cycles. DO NOT FREEZE.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.