Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM+IgG+IgA Goat anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ503MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ503MI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ503MI Supplier MilliporeSigma Supplier No. AQ503
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Goat anti-Human IgM+IgG+IgA Antibody, F(ab′)2 is validated for use in ELISA for the detection of Human IgM+IgG+IgA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Human IgM IgG IgA
Applications ELISA
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Purified by gel filtration chromatography of pepsin digested antibody. Liquid in 1.0mLof 100mM borate buffered saline, pH 8.2.
Host Species Goat
Immunogen pepsin digest of human Ig.
Purification Method Gel filtration chromatography
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C under sterile conditions for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.