Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human Kappa chains, Polyclonal, MP Biomedicals

Catalog No. ICN55059 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55059 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55059 Supplier MP Biomedicals, Inc Supplier No. 0855059
Add to Cart
Add to Cart

Host Species: Goat
Species Reactivity: Human
Conjugate: Unlabeled
Form: lyophilized F(ab')2

Immunostaining

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Unconjugated
Quantity 2 mL
Primary or Secondary Secondary
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.