Learn More
Immunology Consultants anti-Human Prealbumin Antibody (Goat) - Affinity Purified
SDP
Catalog No. 502248921
Encompass_Preferred
This affinity‑purified goat antibody targets human Prealbumin (Transthyretin), a transport protein associated with thyroid hormone and retinol‑binding protein delivery. It provides strong specificity and low background in ELISA, Western blotting, and protein‑quantification workflows. GPRE‑80A is well suited for clinical research, metabolic studies, and assays monitoring nutritional status or liver function. Consistent purification ensures reproducible performance across serum, plasma, and purified protein samples.
This item is not returnable. View return policy
Catalog No. 50-224-8921 Supplier Immunology Consultants Supplier No. GPRE80A
This affinity‑purified goat antibody targets human Prealbumin (Transthyretin), a transport protein associated with thyroid hormone and retinol‑binding protein delivery. It provides strong specificity and low background in ELISA, Western blotting, and protein‑quantification workflows. GPRE‑80A is well suited for clinical research, metabolic studies, and assays monitoring nutritional status or liver function. Consistent purification ensures reproducible performance across serum, plasma, and purified protein samples.