Learn More
Seracare Life Sciences Inc GOAT ANTI-LISTERIA SPECIESHIGH
Supplier:  Seracare Life Sciences Inc 53100320
Encompass_Preferred
Affinity purified BacTrace antibody to the Listeria genus made in goat. Isolated from a serum pool of goats immunized with different strains of heat-killed whole cells of Listeria.
Catalog No. 50-668-58