Learn More
Jackson Immuno Research Labs GOAT ANTI M IGG (H+L)
SDP
Supplier:  Jackson Immuno Research Labs 115006062
Encompass_Preferred
AffiniPure F(ab')2 Fragment Goat Anti-Mouse IgG (H+L) (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC9600907
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov