Learn More
Jackson Immuno Research Labs Alexa Fluor 647 AffiniPure Goat Anti-Mouse IgG, F(ab')₂ fragment specific, 1mg
SDP
Supplier:  Jackson Immuno Research Labs 115605072
Encompass_Preferred
Alexa Fluor 647-AffiniPure Goat Anti-Mouse IgG, F(ab')2 Fragment Specific (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC0608493