Learn More
Immunology Consultants Affinity Purified Goat anti-Mouse C3 Antibody, 0.1 mg
SDP
Supplier:  Immunology Consultants GC390AZ
Encompass_Preferred
Goats were immunized with highly purified mouse C3 and the resulting antiserum was collected. Antibody was immunoaffinity purified off an C3 containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with mouse C3 as determined by IEP and ELISA techniques.It is suitable for blotting, ELISA, and IHC applications.This antibody may cross react with C3 from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8790