Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ig kappa chain Goat anti-Mouse, Polyclonal, Secondary Antibody

Catalog No. AQ500MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ500MI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ500MI Supplier MilliporeSigma™ Supplier No. AQ500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Goat anti-Mouse Ig kappa chain Antibody, F(ab′)2 detects level of Mouse Ig kappa chain and has been published and validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse Ig kappa chain
Applications ELISA
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Purified by gel filtration chromatography of pepsin digested antibody. Liquid in 1.0mLof 100mM borate buffered saline, pH 8.2.
Host Species Goat
Immunogen pepsin digest of mouse kappa light chains.
Purification Method Gel filtration chromatography
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.