Learn More
Invitrogen™ Goat anti-Mouse IgA Cross-Adsorbed Secondary Antibody, HRP
Shop All Thermo Scientific ProductsDescription
This antibody recognizes Mouse and Rat IgA and has been cross absorbed against Mouse IgM, IgG1, IgG2a, IgG2b and IgG3. This antibody will not react with Human samples.
Specifications
Specifications
| Antigen | Mouse IgA Cross-Adsorbed |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.4 |
| Host Species | Goat |
| Immunogen | Mouse IgA paraproteins. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.