Learn More
Description
Goats were immunized with highly purified mouse serum IgA in Freunds adjuvant to produce PA1-29741.
Specifications
Specifications
| Antigen | Mouse IgA |
| Applications | ELISA, Immunoprecipitation, Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Goats were immunized with highly purified mouse serum IgA in Freunds adjuvant. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 500 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.