Learn More
Description
Goat anti mouse IgE polyclonal antibody recognizes mouse IgE and shows negligible cross - reactivity with other immunoglobulins.
Specifications
Specifications
| Antigen | Mouse IgE |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 0.2% BSA and 0.05% ProClin 300 |
| Host Species | Goat |
| Immunogen | Native mouse IgE. |
| Purification Method | Affinity chromatography |
| Quantity | 500 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.