Learn More
Jackson Immuno Research Labs AffiniPure Goat Anti-Mouse IgG, Fcγ fragment specific
SDP
Catalog No. NC9254315
Encompass_Preferred
AffiniPure Goat Anti-Mouse IgG, Fc Fragment Specific (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC9254315 Supplier Jackson Immuno Research Labs Supplier No. 115005071
AffiniPure Goat Anti-Mouse IgG, Fc Fragment Specific (min X Hu,Bov,Hrs Sr Prot)