Learn More
GenDEPOT Goat anti-Mouse IgG
Supplier:  GenDEPOT SA001500
Encompass_Preferred
Affinity purified antibody isolated from a pool of serum from goats immunized with purified mouse IgG was labeled with peroxidase by the periodate method of Nakane and Kawaoi.
Catalog No. 50-190-4205