Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Mouse IgG, Alkaline Phosphatase, Secondary Antibody,

Catalog No. DC05L200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
Conjugate:
Alkaline Phosphatase (AP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DC05L200UG 200 μg Alkaline Phosphatase (AP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DC05L200UG Supplier MilliporeSigma™ Supplier No. DC05L200UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects IgG Clone: in Mouse samples, and it is validated for ELISA, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications ELISA, Immunoblot, Immunoblot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Lyophilized from 250mM NaCl, 10mM Tris-HCl, 15mg/mL BSA, pH 8.0.
Host Species Goat
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2°C to 8°C
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.